Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|
About This Item
Empirical Formula (Hill Notation):
C9H13N3O5
CAS Number:
Molecular Weight:
243.22
UNSPSC Code:
12352200
PubChem Substance ID:
MDL number:
Pricing and availability is not currently available.
form
aqueous solution
extent of labeling
40-60 mCi per mmol
shipped in
dry ice
storage temp.
−20°C
SMILES string
NC1=NC(=O)N(C=C1)C2OC(CO)C(O)C2O
Packaging
serum bottle
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.