Key Documents
BMSIPD960015ZOB
Sigma-Aldrich
BeadBlaster 96 Zirconium Oxide Grinding Ball
bead diam. 15 mm, Zirconium Oxide Grinding grinding jar
Select a Size
2 EACH
CZK 1,830.00
CZK 1,830.00
Please contact Customer Service for Availability
Select a Size
Change View
2 EACH
CZK 1,830.00
About This Item
UNSPSC Code:
41100000
NACRES:
NA.21
CZK 1,830.00
Please contact Customer Service for Availability
material
(Zirconium Oxide Grinding)
packaging
pack of 2
manufacturer/tradename
Benchmark Scientific IPD9600-15ZOB
bead diam.
15 mm
compatibility
Zirconium Oxide Grinding grinding jar
General description
Zirconium Oxide is the most durable and efficient media for ball milling and attrition milling of ceramic materials. Extremely low abrasive wear prevents materials from being polluted. Suitable for the environment of high viscosity, wet grinding and dispersion. It shows high grinding efficiency and long-time working life. Zirconia grinding media provides a virtually contamination-free ball milling solution for a variety of industries.
Application
Homogenization of metallurgy, geology and pharmaceutical samples
Features and Benefits
  • virtually contamination
  • free ball milling
  • High hardness for wear resistance
  • Not corroded by exposure to solvents
Legal Information
BeadBlaster is a trademark of Benchmark Scientific, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)