Skip to Content
Merck

Skip To

B9901

Brain Natriuretic Peptide-32 rat

≥97% (HPLC), powder

Synonym(s):

BNP-32

Sign In to View Organizational & Contract Pricing.

About This Item

Empirical Formula (Hill Notation):
C146H239N47O44S3
CAS Number:
Molecular Weight:
3452.94
UNSPSC Code:
12352200
PubChem Substance ID:
MDL number:
Assay:
≥97% (HPLC)
Form:
powder


assay

≥97% (HPLC)

form

powder

composition

Peptide content, ≥65%

UniProt accession no.

storage temp.

−20°C

SMILES string

CC[C@H](C)[C@@H]1NC(=O)[C@H](CCCNC(N)=N)NC(=O)[C@H](CC(O)=O)NC(=O)[C@@H](NC(=O)[C@H](CCCCN)NC(=O)[C@H](CCC(N)=O)NC(=O)CNC(=O)[C@H](Cc2ccccc2)NC(=O)[C@H](CSSC[C@H](NC(=O)CNC(=O)[C@H](CC(C)C)NC(=O)[C@H](CCCNC(N)=N)NC(=O)[C@H](CO)NC(=O)[C@@H](NC(=O)[C@H](C)NC(=O)CNC1=O)C(C)C)C(=O)N[C@@H](CC(O)=O)C(=O)NCC(=O)N[C@@H](CC(C)C)C(=O)N[C@@H](CCCNC(N)=N)C(=O)N[C@@H](CC(C)C)C(=O)N[C@@H](Cc3ccccc3)C(O)=O)NC(=O)[C@H](CO)NC(=O)[C@H](CO)NC(=O)[C@H](CO)NC(=O)[C@H](Cc4c[nH]cn4)NC(=O)[C@H](C)NC(=O)[C@H](CCSC)NC(=O)[C@H](CCCCN)NC(=O)[C@H](CO)NC(=O)[C@@H](N)CC(N)=O)[C@@H](C)CC

InChI

1S/C146H239N47O44S3/c1-16-75(11)114-140(233)165-59-107(201)167-77(13)117(210)191-113(74(9)10)141(234)189-102(67-198)134(227)176-86(38-29-46-160-145(154)155)124(217)179-90(49-71(3)4)119(212)162-62-110(204)171-103(138(231)182-95(56-111(205)206)121(214)164-61-109(203)170-91(50-72(5)6)129(222)173-85(37-28-45-159-144(152)153)125(218)180-92(51-73(7)8)130(223)184-97(143(236)237)53-80-33-22-19-23-34-80)68-239-240-69-104(139(232)181-93(52-79-31-20-18-21-32-79)120(213)163-60-108(202)169-88(40-41-105(150)199)126(219)172-84(36-25-27-44-148)127(220)193-115(76(12)17-2)142(235)183-96(57-112(207)208)132(225)174-87(128(221)192-114)39-30-47-161-146(156)157)190-137(230)101(66-197)188-136(229)100(65-196)187-135(228)99(64-195)186-131(224)94(54-81-58-158-70-166-81)178-116(209)78(14)168-122(215)89(42-48-238-15)177-123(216)83(35-24-26-43-147)175-133(226)98(63-194)185-118(211)82(149)55-106(151)200/h18-23,31-34,58,70-78,82-104,113-115,194-198H,16-17,24-30,35-57,59-69,147-149H2,1-15H3,(H2,150,199)(H2,151,200)(H,158,166)(H,162,212)(H,163,213)(H,164,214)(H,165,233)(H,167,201)(H,168,215)(H,169,202)(H,170,203)(H,171,204)(H,172,219)(H,173,222)(H,174,225)(H,175,226)(H,176,227)(H,177,216)(H,178,209)(H,179,217)(H,180,218)(H,181,232)(H,182,231)(H,183,235)(H,184,223)(H,185,211)(H,186,224)(H,187,228)(H,188,229)(H,189,234)(H,190,230)(H,191,210)(H,192,221)(H,193,220)(H,205,206)(H,207,208)(H,236,237)(H4,152,153,159)(H4,154,155,160)(H4,156,157,161)/t75-,76-,77-,78-,82-,83-,84-,85-,86-,87-,88-,89-,90-,91-,92-,93-,94-,95-,96-,97-,98-,99-,100-,101-,102-,103-,104-,113-,114-,115-/m0/s1

InChI key

CKSKSHHAEYGHSB-LBCJMRPYSA-N

Gene Information

rat ... Nppb(25105)

Biochem/physiol Actions

Brain natriuretic peptide (type B natriuretic peptide) was originally isolated from brain, but is mainly produced in myoendocrine cells of the heart ventricles from which it is released into the circulation. It is involved in blood pressure control and cardiovascular homeostasis.
Involved in blood pressure control and cardiovascular homeostasis.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters