Skip to Content
Merck

Skip To

B6805

Nalgene® dropper bottle with screw closure

capacity 4 mL, white bottle, assorted colors cap

Synonym(s):

bottle dropper, dropper bottles, Dropper container, Dropper flask, Nalgene 2752-9125, dispensing bottle

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
25 ea
Please contact Customer Service for Availability
CZK 1,360.00
CZK 1,224.00

About This Item

NACRES:
NB.17
UNSPSC Code:
41121800

CZK 1,224.00

List PriceCZK 1,360.00Save 10%

Please contact Customer Service for Availability

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


material

low-density polyethylene , round bottle, white bottle

joint

Joints threaded neck (15-415)

packaging

pack of 25 ea

manufacturer/tradename

Nalgene® 2752-9125

capacity

4 mL

color

assorted colors cap

Features and Benefits

  • Nalgene® dropper bottles with screw closure is an alternative to various pipetting devices.
  • The tip ensures accurate dispensing of 40μL drops (based on water; viscosity affects drop size), dispensed one drop at a time.
  • Chemically resistant to most reagents.
  • LDPE clear transparent bottles for easy sample visualization.
  • Leak-proof closure - prevents sample loss.
  • Control dispensing tip ensures a secure fit.
  • Suitable for use in biotech, diagnostic and pharmaceutical applications.

Legal Information

Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
B6430B6055B6555
manufacturer/tradename

Nalgene® 2752-9125

manufacturer/tradename

Nalgene® 2751-9125

manufacturer/tradename

Nalgene® 2750-9125

manufacturer/tradename

Nalgene® 2751-9025

capacity

4 mL

capacity

4 mL

capacity

4 mL

capacity

8 mL

material

low-density polyethylene , white bottle, round bottle

material

low-density polyethylene , round bottle, white bottle

material

colorless bottle, low-density polyethylene , polypropylene bottle, round bottle

material

low-density polyethylene , round bottle, white bottle

packaging

pack of 25 ea

packaging

pack of 25 ea

packaging

pack of 25 ea

packaging

pack of 25 ea

color

assorted colors cap

color

white cap

color

white cap

color

white cap

joint

Joints threaded neck (15-415)

joint

Joints threaded neck (15-415)

joint

Joints threaded neck (15)

joint

Joints threaded neck (15-415)


Still not finding the right product?




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library