Skip to Content
Merck

Skip To

A9791

Asialoglycophorin from Human Blood Type MN

lyophilized powder

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
1 mg
Please contact Customer Service for Availability
CZK 36,900.00

About This Item

UNSPSC Code:
12352204
NACRES:
NA.54
MDL number:
Biological source:
human blood (type MN)
Concentration:
30.00-50.00% (Lowry)

CZK 36,900.00


Please contact Customer Service for Availability

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

human blood (type MN)

Quality Segment

form

lyophilized powder

concentration

30.00-50.00% (Lowry)

storage temp.

−20°C

Application

Glycophorin is the major sialylated protein of the erythrocyte membrane. Product A9791 is asialoglycophorin from human blood type MN. Glycophorin has been inserted into target cells, by electropulsation, to study resistance to natural killer cells [1].

Biochem/physiol Actions

M and N active glycopeptides have identical amino acid sequences, except at residues 1 and 5, and glycosylated residues 2 to 4, which contain approximately 5 sialic acid groups. The removal of sialic acid from glycophorin destroys M and N activity. Glycophorin A may be involved in the resistance of target cells to natural killing.

Other Notes

Predominantly asialoglycophorin A

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
G5017H2500A5843
biological source

human blood (type MN)

biological source

human blood (type MN)

biological source

bovine blood

biological source

human

concentration

30.00-50.00% (Lowry)

concentration

-

concentration

-

concentration

-

form

lyophilized powder

form

lyophilized powder

form

lyophilized powder

form

lyophilized powder

storage temp.

−20°C

storage temp.

−20°C

storage temp.

2-8°C

storage temp.

2-8°C

Quality Level

200

Quality Level

200

Quality Level

300

Quality Level

200


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters