Skip to Content
Merck

Skip To

A8724

ADP-ribosyltransferase C3 from Clostridium botulinum

Synonym(s):

Botulinum neurotoxin C3, C3 Exoenzyme, C3 Exotoxin, C3 Transferase

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

CAS Number:
UNSPSC Code:
12352204
MDL number:
Product A8724 is not currently sold in your country. Contact Technical Service
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

powder

Quality Level

storage temp.

2-8°C

Application

ADP-ribosyltransferase C3 from Clostridium botulinum may be used to study cellular signaling and G protein expression [1].

Biochem/physiol Actions

ADP-ribosyltransferase C3 from Clostridium botulinum ribosylates rho in eukaryotes in the presence of NAD.[1] The ADP-ribosylating exoenzyme forms a single major 25 kDA (approx.) band with SDS electrophoresis. The enzyme labels 21-24 kDa proteins in tissues such as the human platelet membranes.[2]
Ribosylates rho in eukaryotes in the presence of NAD.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Jordi Farrés et al.
Blood, 122(1), 44-54 (2013-05-17)
Hematopoietic stem cells self-renew for life to guarantee the continuous supply of all blood cell lineages. Here we show that Poly(ADP-ribose) polymerase-2 (Parp-2) plays an essential role in hematopoietic stem/progenitor cells (HSPC) survival under steady-state conditions and in response to
J Michels et al.
Oncogene, 33(30), 3894-3907 (2013-09-17)
Poly(ADP-ribose) polymerase (PARP) inhibitors have raised high expectations for the treatment of multiple malignancies. PARP inhibitors, which can be used as monotherapies or in combination with DNA-damaging agents, are particularly efficient against tumors with defects in DNA repair mechanisms, in
Françoise Dantzer et al.
The FEBS journal, 280(15), 3508-3518 (2013-06-05)
Poly(ADP-ribose) polymerases (PARPs) are enzymes that transfer poly(ADP-ribose) (PAR) groups to target proteins, and thereby affect various nuclear and cytoplasmic processes. The activity of PARP family members, such as PARP1 and PARP2, is tied to cellular signalling pathways, and, through



Global Trade Item Number

SKUGTIN
A8724-50UG04061832921273

Questions

Reviews

No rating value

Active Filters