Skip to Content
Merck

Skip To

A7980

AC-41848 hydrate

≥98% (HPLC), solid

Synonym(s):

1-[(2,4-Dichlorophenyl)methyl]-6,7,8,9-tetrahydro-3-phenyl-5H-imidazo[1,2-a]azepinium bromide hydrate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
20 mg
Please contact Customer Service for Availability
11 900,00 CZK

About This Item

Empirical Formula (Hill Notation):
C21H21BrCl2N2 · xH2O
CAS Number:
Molecular Weight:
452.21 (anhydrous basis)
NACRES:
NA.77
PubChem Substance ID:
UNSPSC Code:
12352200
MDL number:
Assay:
≥98% (HPLC)
Form:
solid
Storage condition:
desiccated, under inert gas

11 900,00 CZK


Please contact Customer Service for Availability

Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

solid

storage condition

desiccated, under inert gas

solubility

DMSO: ~14 mg/mL, H2O: >5 mg/mL

storage temp.

2-8°C

SMILES string

[Br-].[H]O[H].Clc1ccc(C[n+]2cc(-c3ccccc3)n4CCCCCc24)c(Cl)c1

InChI

1S/C21H21Cl2N2.BrH.H2O/c22-18-11-10-17(19(23)13-18)14-24-15-20(16-7-3-1-4-8-16)25-12-6-2-5-9-21(24)25;;/h1,3-4,7-8,10-11,13,15H,2,5-6,9,12,14H2;1H;1H2/q+1;;/p-1

InChI key

FPOBEBBUBSFIIA-UHFFFAOYSA-M

Application

AC-41848 is a potent, cell permeable, and subtype selective retinoic acid receptor RARγ agonist.

Biochem/physiol Actions

AC-41848 is a potent, cell permeable, subtype selective retinoic acid receptor RARγ agonist. AC-41848 has high selectivity (92%) for RARγ. EC50 = 5.9 μM.
Potent, cell permeable, subtype selective retinoic acid receptor RARγ agonist. High selectivity (92%) for RARγ. EC50 = 5.9 μM.

Features and Benefits

This compound is featured on the Nuclear Receptors (Non-Steroids) page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
A9736SML0178B3686
description

≥98% (HPLC), solid

description

≥98% (HPLC)

description

≥98% (HPLC)

description

solid, ≥98% (HPLC)

form

solid

form

powder

form

powder

form

solid

assay

≥98% (HPLC)

assay

≥98% (HPLC)

assay

≥98% (HPLC)

assay

≥98% (HPLC)

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

storage condition

desiccated, under inert gas

storage condition

desiccated

storage condition

desiccated

storage condition

protect from light

solubility

H2O: >5 mg/mL, DMSO: ~14 mg/mL

solubility

DMSO: 20 mg/mL, clear

solubility

H2O: ≥5 mg/mL

solubility

H2O: >8 mg/mL


pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters