Skip to Content
Merck

Skip To

A5625

N-Acetyl-DL-aspartic acid

≥95%

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice
5 g
Please contact Customer Service for Availability
CZK 7,110.00

About This Item

Empirical Formula (Hill Notation):
C6H9NO5
CAS Number:
Molecular Weight:
175.14
UNSPSC Code:
12352209
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.26
EC Number:
219-827-5
MDL number:

CZK 7,110.00


Please contact Customer Service for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

N-Acetyl-DL-aspartic acid, ≥95%

Quality Level

assay

≥95%

form

powder

color

white to off-white

storage temp.

2-8°C

SMILES string

CC(=O)NC(CC(O)=O)C(O)=O

InChI

1S/C6H9NO5/c1-3(8)7-4(6(11)12)2-5(9)10/h4H,2H2,1H3,(H,7,8)(H,9,10)(H,11,12)

InChI key

OTCCIMWXFLJLIA-UHFFFAOYSA-N


Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
A0637A9006H4398
form

powder

form

powder

form

powder

form

powder

assay

≥95%

assay

≥99%

assay

≥99% (TLC)

assay

≥98%

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

-

storage temp.

2-8°C

color

white to off-white

color

-

color

white

color

-

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

100


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



L Boughamoura et al.
Archives de pediatrie : organe officiel de la Societe francaise de pediatrie, 14(2), 173-176 (2007-01-02)
Canavan disease or N-acetyl aspartic aciduria, is an autosomal recessive leukodystrophy characterized by spongy degeneration of brain. The disease is an inborn error of metabolism caused by aspartoacylase deficiency resulting from accumulation of N-acetyl aspartic acid in the brain. The
Zhongyu Hou et al.
Journal of clinical neuroscience : official journal of the Neurosurgical Society of Australasia, 17(5), 628-633 (2010-02-02)
In recent years, investigations of the pathologic mechanism of Parkinson's disease (PD) have mainly concentrated on the basal ganglia. However, recent studies have confirmed that pathological changes in PD are accompanied by functional motor changes of the cerebral cortex. Rats
J A Harvey et al.
The Biochemical journal, 108(2), 269-274 (1968-06-01)
1. Sample from the neocortex and piriform cortex of guinea pigs and rats were incubated in inulin-containing glucose-saline. Their intracellular (non-inulin) space contained 19-27muequiv. of Na(+)/g. of original tissue. These values were stable between 30 and 100min. after incubation commenced



Global Trade Item Number

SKUGTIN
A5625-5G04061832421155

Questions

Reviews

No rating value

Active Filters