product line
BioReagent
form
solid
manufacturer/tradename
ATTO-TEC GmbH
fluorescence
λex 535 nm; λem 560 nm
suitability
suitable for fluorescence
storage temp.
−20°C
General description
Atto Rho6G-Biotin is a new rhodamine dye, based on the well-known laser dye Rhodamine 6G. The new label is functionalized for coupling to biomolecules such as DNA, RNA or proteins. Atto Rho6G-Biotin shows strong absorption, extraordinary high fluorescence quantum yield, high thermal and photo-stability, and very little triplet formation.
After coupling to a substrate Atto Rho6G-Biotin carries a net electric charge of +1.
The label is moderately hydrophilic.
find more information here
After coupling to a substrate Atto Rho6G-Biotin carries a net electric charge of +1.
The label is moderately hydrophilic.
find more information here
1 of 1
This Item | |||
|---|---|---|---|
| suitability suitable for fluorescence | suitability suitable for fluorescence | suitability suitable for fluorescence | suitability - |
| fluorescence λex 535 nm; λem 560 nm | fluorescence λex 535 nm; λem 560 nm | fluorescence - | fluorescence - |
| form solid | form solid | form solid | form solid |
| storage temp. −20°C | storage temp. −20°C | storage temp. −20°C | storage temp. −20°C |
| product line BioReagent | product line BioReagent | product line - | product line BioReagent |
| manufacturer/tradename ATTO-TEC GmbH | manufacturer/tradename ATTO-TEC GmbH | manufacturer/tradename ATTO-TEC GmbH | manufacturer/tradename ATTO-TEC GmbH |
Storage Class
11 - Combustible Solids
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
