Skip to Content
Merck

Skip To

81062

Atto 514 maleimide

suitable for fluorescence

Sign In to View Organizational & Contract Pricing.

About This Item

NACRES:
NA.32
UNSPSC Code:
12352200


form

solid

Quality Level

mol wt

Mw 990 g/mol

manufacturer/tradename

ATTO-TEC GmbH

fluorescence

λex 511 nm; λem 533 nm±10 nm in PBS, pH 7.4

suitability

suitable for fluorescence

storage temp.

−20°C

General description

Atto 514 is a new hydrophilic fluorescent label with excellent water solubility. The dye exhibits strong absorption, high fluorescence quantum yield and exceptional thermal and photo-stability. Thus Atto 514 is highly suitable for single-molecule detection applications and high-resolution microscopy such as PALM, dSTORM, STED etc. Additionally the dye highly qualifies to be applied in flow cytometry (FACS), fluorescence in-situ hybridization (FISH) and many more. The fluorescence is excited most efficiently in the range 510 - 535 nm. A suitable source of excitation is the 514 nm line of the Argon-Ion laser.
The maleimide is suitable for labeling sulfhydryl (thiol) groups of proteins, in particular cystein residues.

find more information here

Legal Information

This product is for Research use only. In case of intended commercialization, please contact the IP-holder (ATTO-TEC GmbH, Germany) for licensing.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library


Related Content





Questions

Reviews

No rating value

Active Filters