Skip to Content
Merck

Skip To

77085

Atto Rho101

BioReagent, suitable for fluorescence

Sign In to View Organizational & Contract Pricing.

About This Item

UNSPSC Code:
12352200


product line

BioReagent

form

solid

manufacturer/tradename

ATTO-TEC GmbH

fluorescence

λex 586 nm; λem 610 nm

suitability

suitable for fluorescence

storage temp.

−20°C

General description

Atto Rho101 is a new rhodamine dye, based on the well-known laser dye Rhodamine 101. The new label is functionalized for coupling to bio-molecules such as DNA, RNA or proteins. Atto Rho101 shows extraordinary brightness and photostability.
After coupling to a substrate Atto Rho101 carries a net electrical charge of +1. The dye is moderately hydrophilic

Legal Information

This product is for Research use only. In case of intended commercialization, please contact the IP-holder (ATTO-TEC GmbH, Germany) for licensing.


Storage Class

13 - Non Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library






Global Trade Item Number

SKUGTIN
77085-1MG04061823369855

Questions

Reviews

No rating value

Active Filters