Skip to Content
Merck

Skip To

68841

Dextran sulfate sodium salt from Leuconostoc spp.

Mr ~20,000

Sign In to View Organizational & Contract Pricing.

About This Item

CAS Number:
UNSPSC Code:
12352201
NACRES:
NA.25
MDL number:


form

powder

mol wt

Mr ~20,000

composition

Sulfur Content, 16.0-20.0%

loss

≤10% loss on drying

solubility

H2O: 100 mg/mL, clear, colorless to faintly greenish-yellow

storage temp.

room temp

General description

Dextran is a polymer of anhydroglucose. It is composed of approximately 95% alpha-D-(166) linkages. The remaining (163) linkages account for the branching of dextran.

Application

Dextran sulfate is routinely used for the selective precipitation of lipoproteins, probe hybridization membrane-immobilized DNA, the release of DNA from DNA-histone complexes, and the inhibition of RNA binding to ribosomes.

Biochem/physiol Actions

Dextran sulfate can interact and bind to lipids making it useful in protein isolation techniques to remove contaminating lipid molecules. [1]

Preparation Note

Sigma dextrans are derived from Leuconostoc mesenteroides.

Other Notes

To gain a comprehensive understanding of our extensive range of Dextrans for your research, we encourage you to visit our Carbohydrates Category page.


Still not finding the right product?


Storage Class

11 - Combustible Solids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library


Articles

Prozkoumejte polysacharidy značené FITC a jejich vlastnosti, včetně vlivu molekulové hmotnosti na rozměry molekul.

Explore FITC-labelled polysaccharides and their properties, including impact of molecular weight on molecule dimensions.


H-D Bae et al.
Poultry science, 88(8), 1703-1711 (2009-07-11)
In avian species, blood IgY is selectively incorporated into the yolks of maturing oocytes, although the precise mechanism is poorly understood. Our previous study showed that 22% of i.v.-injected heterologous chicken IgY (cIgY) was incorporated into egg yolks of Japanese
Aaron H Rose et al.
Inflammatory bowel diseases, 21(9), 2005-2015 (2015-06-16)
An important role has emerged for calpain enzymes in regulating inflammation with one isoform, calpain-2, particularly important for macrophage activation. The goal of this study was to determine the therapeutic potential of a synthetic calpain-2 inhibitor, zLLY-CH2F, for colitis and



Global Trade Item Number

SKUGTIN
68841-100G04061833031230
68841-5G04061833031254
68841-25G04061833031247

Questions

Reviews

No rating value

Active Filters