Skip to Content
Merck
All Photos(1)

Key Documents

43307

Sigma-Aldrich

1,2-Distearoyl-sn-glycero-3-phospho-L-serine sodium salt

≥75% (TLC)

Synonym(s):

(2S,8R)-2-Amino-5-hydroxy-11-oxo-8-[(1-oxooctadecyl)oxy]-4,6,10-trioxa-5-phosphaoctacosanoic acid 5-oxide sodium salt, 1,2-Dioctadecanoyl-sn-glycero-3-phosphoserine sodium salt, 1,2-Dioctadecanoyl-sn-glycero-3-phospho-L-serine sodium salt, 3-sn-Phosphatidyl-L-serine, distearoyl sodium salt, DSPS-Na, PS(18:0/18:0)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C42H81NNaO10P
CAS Number:
Molecular Weight:
814.06
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.85

biological source

synthetic

Assay

≥75% (TLC)

form

crystalline

functional group

carboxylic acid

lipid type

phosphoglycerides

storage temp.

−20°C

SMILES string

[Na+].CCCCCCCCCCCCCCCCCC(=O)OC[C@H](COP([O-])(=O)OC[C@H](N)C(O)=O)OC(=O)CCCCCCCCCCCCCCCCC

InChI

1S/C42H82NO10P.Na/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-31-33-40(44)50-35-38(36-51-54(48,49)52-37-39(43)42(46)47)53-41(45)34-32-30-28-26-24-22-20-18-16-14-12-10-8-6-4-2;/h38-39H,3-37,43H2,1-2H3,(H,46,47)(H,48,49);/q;+1/p-1/t38-,39+;/m1./s1

InChI key

SJRBMGBLPUBGJL-MBAWARMDSA-M

Application

In a study of phase transitions in cholesterol-phospholipid mixtures, cholesterol was found to be much less miscible with DSPS in the gel phase than the liquid crystal phase; up to 50 mol % cholesterol fails to abolish the hydrocarbon chain-melting phase transition.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Filippo Caschera et al.
PloS one, 5(1), e8546-e8546 (2010-01-06)
We consider the problem of optimizing a liposomal drug formulation: a complex chemical system with many components (e.g., elements of a lipid library) that interact nonlinearly and synergistically in ways that cannot be predicted from first principles. The optimization criterion
Kouhei Ohya et al.
Antimicrobial agents and chemotherapy, 63(4) (2019-02-06)
The present study aimed to clarify the mechanism underlying the high distribution of lascufloxacin in epithelial lining fluid (ELF). Involvement of transporters was examined by transcellular transport across Calu-3 and transporter-overexpressing cells; the binding of lascufloxacin to ELF components was
Jing Song et al.
Journal of applied biomaterials & functional materials, 18, 2280800020942712-2280800020942712 (2020-11-06)
A neurodegenerative disorder, glaucoma is a leading cause of blindness in the world. The conventional treatment strategies do not allow the significant penetration of the drug in the cornea. Therefore, we prepare a brinzolamide (Brz) loaded core-shell nanoparticles (NPs) to
Richard H Carson et al.
Journal of lipid research, 58(9), 1884-1892 (2017-07-27)
Compartmentalization of metabolism into specific regions of the cell, tissue, and organ is critical to life for all organisms. Mass spectrometric imaging techniques have been valuable in identifying and quantifying concentrations of metabolites in specific locations of cells and tissues
Stephanie Jemielity et al.
PLoS pathogens, 9(3), e1003232-e1003232 (2013-04-05)
Human T-cell Immunoglobulin and Mucin-domain containing proteins (TIM1, 3, and 4) specifically bind phosphatidylserine (PS). TIM1 has been proposed to serve as a cellular receptor for hepatitis A virus and Ebola virus and as an entry factor for dengue virus.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service