Skip to Content
Merck

Skip To

36035

UV-VIS detectable IEF Marker solution

pI 10.4, 1 mg/mL in 10 mM NaOH, 0.45 μm filtered

Sign In to View Organizational & Contract Pricing.

About This Item

NACRES:
NA.26
UNSPSC Code:
41105336


sterility

0.45 μm filtered

concentration

1 mg/mL in 10 mM NaOH

storage temp.

2-8°C

Application

Suitable as IEF marker for HPCE and IEF gels.


Still not finding the right product?


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
470082-5G04061832425290
36035-200UL04061837837388

Questions

Reviews

No rating value

Active Filters