Skip to Content
Merck

Skip To

19534

Q-Dextran

average mol wt 4,000

Sign In to View Organizational & Contract Pricing.

About This Item

NACRES:
NA.32
UNSPSC Code:
12171500


mol wt

average mol wt 4,000

Quality Segment

composition

nitrogen content, 1.5-2.5%

loss

≤10% loss on drying

storage temp.

−20°C


Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
313947881680881
storage temp.

−20°C

storage temp.

room temp

storage temp.

−20°C

storage temp.

−20°C

mol wt

average mol wt 4,000

mol wt

Mr ~1,500

mol wt

average mol wt 20,000

mol wt

average mol wt 40,000

loss

≤10% loss on drying

loss

-

loss

≤10% loss on drying

loss

≤10% loss on drying

composition

nitrogen content, 1.5-2.5%

composition

-

composition

nitrogen content, 2.5-4.5%

composition

nitrogen content, 2-5%


Still not finding the right product?

Explore all of our products under Q-Dextran


Storage Class

11 - Combustible Solids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library


Articles

Prozkoumejte polysacharidy značené FITC a jejich vlastnosti, včetně vlivu molekulové hmotnosti na rozměry molekul.

Explore FITC-labelled polysaccharides and their properties, including impact of molecular weight on molecule dimensions.






Global Trade Item Number

SKUGTIN
19534-1G04061826567395

Questions

Reviews

No rating value

Active Filters