Skip to Content
Merck

Skip To

14652

D-Mannose 6-sulfate sodium salt

≥90% (TLC)

Synonym(s):

Man-6S

Sign In to View Organizational & Contract Pricing.

About This Item

Empirical Formula (Hill Notation):
C6H11NaO9S
CAS Number:
Molecular Weight:
282.20
UNSPSC Code:
12352201
PubChem Substance ID:
MDL number:
Assay:
≥90% (TLC)


assay

≥90% (TLC)

storage temp.

−20°C

SMILES string

[Na+].OC1O[C@H](COS([O-])(=O)=O)[C@@H](O)[C@H](O)[C@@H]1O

InChI

1S/C6H12O9S.Na/c7-3-2(1-14-16(11,12)13)15-6(10)5(9)4(3)8;/h2-10H,1H2,(H,11,12,13);/q;+1/p-1/t2-,3-,4+,5+,6?;/m1./s1

InChI key

MKFZWUBBFWXXKZ-XXIBIAAKSA-M

Application

D-Mannose 6-sulfate (Man-6S) occurs in some N-glycans that occur on proteins such as Dictyostelium discoideum cathepsin D and various lysosomal enzymes. Mannose 6-sulfate can be used as a reference material in the chemical analysis of Man-6S glycans.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.


Storage Class

13 - Non Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
359130-5ML04061838047502

Questions

Reviews

No rating value

Active Filters