Skip to Content
Merck

Skip To

RDEN1502

Relative Density Standard (15°C)

UKAS ISO/IEC17025 and ISO 17034 certified, Relative Density 15/4 0.7186 at 15 °C

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
500 mL
Please contact Customer Service for Availability
CZK 16,500.00

About This Item

NACRES:
NA.24
UNSPSC Code:
41116107

CZK 16,500.00


Please contact Customer Service for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

certified reference material

Quality Segment

shelf life

limited shelf life, expiry date on the label

packaging

pkg of 500 mL

manufacturer/tradename

Paragon Scientific Ltd

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

storage temp.

2-8°C

General description

This product is a certified reference material (CRM) certified to International Standards BS EN ISO / IEC 17025 and ISO 17034 under UKAS accreditation

Paragon ISO 17025 / ISO 17034 certified Relative Density standards are for the verification of instruments used to measure density of materials which are fluid at the desired test temperature within the range of 15°C to 150°C.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
RDEN1503RDEN1504RDEN1506
grade

certified reference material

grade

certified reference material

grade

certified reference material

grade

certified reference material

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

manufacturer/tradename

Paragon Scientific Ltd

manufacturer/tradename

Paragon Scientific Ltd

manufacturer/tradename

Paragon Scientific Ltd

manufacturer/tradename

Paragon Scientific Ltd

shelf life

limited shelf life, expiry date on the label

shelf life

limited shelf life, expiry date on the label

shelf life

limited shelf life, expiry date on the label

shelf life

limited shelf life, expiry date on the label

packaging

pkg of 500 mL

packaging

pkg of 500 mL

packaging

pkg of 500 mL

packaging

pkg of 500 mL

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100


Still not finding the right product?

Explore all of our products under Relative Density Standard (15°C)


signalword

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Asp. Tox. 1 - Carc. 1B - Flam. Liq. 2

Storage Class

3 - Flammable liquids

wgk

WGK 3

flash_point_f

-4.0 °F

flash_point_c

-20 °C


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

64742-53-6

CAS No.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters