About This Item
grade
analytical standard
Quality Level
form
solid
technique(s)
HPLC: suitable, gas chromatography (GC): suitable
color
off-white
solubility
H2O: soluble, ethanol: soluble
application(s)
pharmaceutical (small molecule)
format
neat
SMILES string
OC(=O)\C=C/C(O)=O.CC(O)c1cccc2cc(oc12)C(O)CNC(C)(C)C
InChI
1S/C16H23NO3.C4H4O4/c1-10(18)12-7-5-6-11-8-14(20-15(11)12)13(19)9-17-16(2,3)4;5-3(6)1-2-4(7)8/h5-8,10,13,17-19H,9H2,1-4H3;1-2H,(H,5,6)(H,7,8)/b;2-1-
InChI key
UCHVLIWUAUXBBC-BTJKTKAUSA-N
Application
Biochem/physiol Actions
1 of 1
This Item | |||
|---|---|---|---|
| grade analytical standard | grade analytical standard | grade analytical standard | grade analytical standard |
| format neat | format neat | format neat | format neat |
| technique(s) HPLC: suitable, gas chromatography (GC): suitable | technique(s) HPLC: suitable, gas chromatography (GC): suitable | technique(s) HPLC: suitable, gas chromatography (GC): suitable | technique(s) HPLC: suitable, gas chromatography (GC): suitable |
| Quality Level 200 | Quality Level 200 | Quality Level 200 | Quality Level 100 |
| solubility H2O: soluble, ethanol: soluble | solubility - | solubility - | solubility - |
| form solid | form - | form - | form - |
Still not finding the right product?
Explore all of our products under Hydroxybufuralol maleate salt
Storage Class
11 - Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Global Trade Item Number
| SKU | GTIN |
|---|---|
| H147-2MG | 04061832850351 |



