Skip to Content
Merck
All Photos(1)

Key Documents

CCW4742

Millipore

TS AGAR

Solid selective and differential medium for isolation and presumptive identification of Clostridium perfringens, suitable for microbiology

Synonym(s):

TS Agar, Tryptone Sulfite Agar

Sign Into View Organizational & Contract Pricing

Select a Size

30 EACH
CZK 2,400.00

CZK 2,400.00


Please contact Customer Service for Availability

Request more information

Select a Size

Change View
30 EACH
CZK 2,400.00

About This Item

UNSPSC Code:
41171606

CZK 2,400.00


Please contact Customer Service for Availability

Request more information

Agency

ISO 11133

Quality Level

form

solid

shelf life

6 mo. (Expiry date on the label)

feature

ready-to-use

composition

agar, 14.00 g/L
enzymatic digest of casein, 15.00 g/L
ferric ammonium citrate, 1.00 g/L
sodium meta-bisulfite, 1.00 g/L
soy Peptone, 5.00 g/L
yeast Extract, 5.00 g/L

packaging

box of 30 plates

storage condition

(Store dry and tightly closed)

parameter

9 mL filling volume

plate diam.

55 mm

color

yellow

pH

7.6±0.2 (25 °C)

application(s)

microbiology

storage temp.

2-25°C

General description

TS Agar is a solid, selective, and differential medium for the isolation and presumptive identification of Clostridium perfringens. The medium does not contain cycloserine. The presence of sodium meta-bisulfite and ferric ammonium citrate allows differential characteristics, such as sulfite reduction and growth at 46°C, of this anaerobic species to be verified with just one assay.

Application

TS Agar plates are the ready-to-use plates for the isolation and presumptive identification of Clostridium perfringens.

Components

1 box containing: 6 plastic bags with 5 plates of 55 mm/bag

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service