Skip to Content
Merck

Skip To

BP714

Norgestrel

British Pharmacopoeia (BP) Reference Standard

Synonym(s):

(±)-Norgestrel, (17α)-(±)-13-Ethyl-17-hydroxy-18,19-dinorpregn-4-en-20-yn-3-one

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice
100 mg
Please contact Customer Service for Availability
CZK 7,520.00

About This Item

Empirical Formula (Hill Notation):
C21H28O2
CAS Number:
Molecular Weight:
312.45
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

CZK 7,520.00


Please contact Customer Service for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

API family

norgestrel

form

solid

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

BP

application(s)

pharmaceutical
pharmaceutical small molecule

format

neat

SMILES string

O[C@@]1([C@@]2([C@H]([C@H]3[C@@H]([C@H]4CCC(=O)C=C4CC3)CC2)CC1)CC)C#C

InChI

1S/C21H28O2/c1-3-20-11-9-17-16-8-6-15(22)13-14(16)5-7-18(17)19(20)10-12-21(20,23)4-2/h2,13,16-19,23H,3,5-12H2,1H3/t16-,17+,18+,19-,20-,21-/m0/s1

InChI key

WWYNJERNGUHSAO-XUDSTZEESA-N

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Norgestrel BP Reference standard, intended for use in laboratory tests only as specifically prescribed in the British Pharmacopoeia.

Also used in monographs such as:
  • Levonorgestrel Tablets
  • Norgestrel Tablets
  • Packaging

    Unit quantity: 100 mg. Subject to change. The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity please visit British Pharmacopoeia

    Other Notes

    Sales restrictions may apply.

    Compare Similar Items

    View Full Comparison

    Show Differences

    1 of 1

    This Item
    N12500001472000PHR2487
    vibrant-m

    BP714

    Norgestrel

    manufacturer/tradename

    BP

    manufacturer/tradename

    EDQM

    manufacturer/tradename

    USP

    manufacturer/tradename

    -

    grade

    pharmaceutical primary standard

    grade

    pharmaceutical primary standard

    grade

    pharmaceutical primary standard

    grade

    certified reference material, pharmaceutical secondary standard

    format

    neat

    format

    neat

    format

    neat

    format

    -

    form

    solid

    form

    -

    form

    -

    form

    solid

    application(s)

    pharmaceutical
    pharmaceutical small molecule

    application(s)

    pharmaceutical (small molecule)

    application(s)

    pharmaceutical (small molecule)

    application(s)

    pharmaceutical small molecule

    shelf life

    limited shelf life, expiry date on the label

    shelf life

    -

    shelf life

    -

    shelf life

    -


    Still not finding the right product?

    Explore all of our products under Norgestrel


    pictograms

    Health hazardExclamation mark

    signalword

    Danger

    Hazard Classifications

    Eye Irrit. 2 - Repr. 1B - Skin Irrit. 2

    Storage Class

    6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

    wgk

    WGK 3

    flash_point_f

    Not applicable

    flash_point_c

    Not applicable



    Choose from one of the most recent versions:

    Certificates of Analysis (COA)

    Lot/Batch Number

    It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

    If you need assistance, please contact Customer Support

    Already Own This Product?

    Find documentation for the products that you have recently purchased in the Document Library.

    Visit the Document Library



    Questions

    Reviews

    No rating value

    Active Filters