Select a Size
| Size/SKU | Availability | Price |
|---|---|---|
100 mg | Please contact Customer Service for Availability | CZK 6,360.00 |
About This Item
grade
pharmaceutical primary standard
API family
methadone
form
solid
shelf life
limited shelf life, expiry date on the label
manufacturer/tradename
BP
drug control
kontrollierte Droge in Deutschland
application(s)
pharmaceutical
pharmaceutical (small molecule)
format
neat
storage temp.
2-8°C
SMILES string
[Cl-].[N+H](C(CC(c2ccccc2)(c1ccccc1)C(=O)CC)C)(C)C
InChI
1S/C21H27NO.ClH/c1-5-20(23)21(16-17(2)22(3)4,18-12-8-6-9-13-18)19-14-10-7-11-15-19;/h6-15,17H,5,16H2,1-4H3;1H
InChI key
FJQXCDYVZAHXNS-UHFFFAOYSA-N
General description
Application
Also used in monographs such as:
Packaging
Other Notes
Legal Information
Dieses Produkt fällt unter das Betäubungsmittelgesetz (BtMG). Für eine Bestellung dieses Produktes ist eine Erlaubnis nach § 3 BtMG zwingend erforderlich, es sei denn, es greift eine Ausnahme von der Erlaubnispflicht nach § 4 oder § 26 BtMG.
English
This product is subject to the German Narcotics Act. A permit under Section 3 of the German Narcotics Act is mandatory for ordering this product unless an exemption from the permit requirement under Section 4 or Section 26 of the German Narcotics Act applies.
1 of 1
This Item | |||
|---|---|---|---|
| grade pharmaceutical primary standard | grade certified reference material | grade pharmaceutical primary standard | grade pharmaceutical primary standard |
| manufacturer/tradename BP | manufacturer/tradename Cerilliant® | manufacturer/tradename BP | manufacturer/tradename BP |
| shelf life limited shelf life, expiry date on the label | shelf life - | shelf life limited shelf life, expiry date on the label | shelf life limited shelf life, expiry date on the label |
| form solid | form liquid | form solid | form powder |
| application(s) pharmaceutical | application(s) forensics and toxicology | application(s) pharmaceutical | application(s) pharmaceutical |
| format neat | format single component solution | format neat | format neat |
Still not finding the right product?
Explore all of our products under Methadone hydrochloride
signalword
Danger
hcodes
pcodes
Hazard Classifications
Acute Tox. 2 Oral - STOT SE 3
target_organs
Central nervous system
Storage Class
6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.



