Skip to Content
Merck

Skip To

BP433

Lithium lactate

British Pharmacopoeia (BP) Reference Standard

Synonym(s):

Lactic acid lithium salt

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice
100 mg
Please contact Customer Service for Availability
CZK 6,250.00

About This Item

Linear Formula:
CH3CH(OH)COOLi
CAS Number:
Molecular Weight:
96.01
EC Number:
212-761-8
UNSPSC Code:
41116107
NACRES:
NA.24
Beilstein/REAXYS Number:
3568332
MDL number:

CZK 6,250.00


Please contact Customer Service for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

API family

lithium lactate

form

crystalline

quality

British Pharmacopoeia (BP) Reference Standard

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

BP

mp

>300 °C (lit.)

application(s)

pharmaceutical
pharmaceutical small molecule

format

neat

storage temp.

2-8°C

SMILES string

[Li+].CC(O)C([O-])=O

InChI

1S/C3H6O3.Li/c1-2(4)3(5)6;/h2,4H,1H3,(H,5,6);/q;+1/p-1

InChI key

GKQWYZBANWAFMQ-UHFFFAOYSA-M

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Lithium lactate BP Reference standard, intended for use in laboratory tests only as specifically prescribed in the British Pharmacopoeia.

Also used in monographs such as:
  • Ciprofloxacin Infusion
  • Compound Sodium Lactate Infusion
  • Sodium Lactate Infusion
  • Packaging

    Unit quantity: 100 mg. Subject to change. The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity please visit British Pharmacopoeia

    Other Notes

    Sales restrictions may apply.

    Compare Similar Items

    View Full Comparison

    Show Differences

    1 of 1

    This Item
    440469517992920320
    manufacturer/tradename

    BP

    manufacturer/tradename

    -

    manufacturer/tradename

    -

    manufacturer/tradename

    -

    grade

    pharmaceutical primary standard

    grade

    -

    grade

    -

    grade

    -

    application(s)

    pharmaceutical
    pharmaceutical small molecule

    application(s)

    -

    application(s)

    -

    application(s)

    battery precursors
    catalysts
    material synthesis precursor

    shelf life

    limited shelf life, expiry date on the label

    shelf life

    -

    shelf life

    -

    shelf life

    -

    format

    neat

    format

    -

    format

    -

    format

    -

    form

    crystalline

    form

    solid

    form

    powder or crystals

    form

    powder


    Still not finding the right product?

    Explore all of our products under Lithium lactate


    Storage Class

    11 - Combustible Solids

    wgk

    WGK 2

    flash_point_f

    Not applicable

    flash_point_c

    Not applicable



    Choose from one of the most recent versions:

    Certificates of Analysis (COA)

    Lot/Batch Number

    It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

    If you need assistance, please contact Customer Support

    Already Own This Product?

    Find documentation for the products that you have recently purchased in the Document Library.

    Visit the Document Library



    Global Trade Item Number

    SKUGTIN
    BP43304061835278046

    Questions

    Reviews

    No rating value

    Active Filters