Skip to Content
Merck

Skip To

93019

Taurine

certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Synonym(s):

2-Aminoethanesulfonic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
100 mg
Please contact Customer Service for Availability
CZK 6,470.00

About This Item

Linear Formula:
NH2CH2CH2SO3H
CAS Number:
Molecular Weight:
125.15
EC Number:
203-483-8
UNSPSC Code:
12352106
NACRES:
NA.24
Beilstein/REAXYS Number:
1751215
MDL number:

CZK 6,470.00


Please contact Customer Service for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

product line

TraceCERT®

grade

certified reference material, TraceCERT®

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

technique(s)

gas chromatography (GC): suitable

mp

>300 °C (lit.)

application(s)

clinical testing

format

neat

SMILES string

NCCS(O)(=O)=O

InChI

1S/C2H7NO3S/c3-1-2-7(4,5)6/h1-3H2,(H,4,5,6)

InChI key

XOAAWQZATWQOTB-UHFFFAOYSA-N

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com.

Biochem/physiol Actions

Non-selective endogenous agonist at glycine receptors.
Non-selective endogenous agonist at glycine receptors. Conditionally essential sulfonated amino acid which modulates apoptosis in some cells; functions in many metabolic activities; a product of methionine and cysteine metabolism.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
PHR11091643361T8691
technique(s)

gas chromatography (GC): suitable

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

technique(s)

-

technique(s)

cell culture | mammalian: suitable

application(s)

clinical testing

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care
pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

application(s)

-

format

neat

format

neat

format

neat

format

-

Quality Level

300

Quality Level

300

Quality Level

-

Quality Level

200

grade

certified reference material, TraceCERT®

grade

certified reference material, pharmaceutical secondary standard

grade

pharmaceutical primary standard

grade

-

shelf life

limited shelf life, expiry date on the label

shelf life

-

shelf life

-

shelf life

-


Still not finding the right product?

Explore all of our products under Taurine


Storage Class

11 - Combustible Solids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters