Skip to Content
Merck

Skip To

75296

Metribuzin

certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Sign In to View Organizational & Contract Pricing.

About This Item

Empirical Formula (Hill Notation):
C8H14N4OS
CAS Number:
Molecular Weight:
214.29
EC Number:
244-209-7
UNSPSC Code:
41116107
NACRES:
NA.24
Beilstein/REAXYS Number:
746650
MDL number:


Quality Segment

product line

TraceCERT®

grade

certified reference material, TraceCERT®

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

format

neat

storage temp.

2-8°C

SMILES string

CSC1=NN=C(C(=O)N1N)C(C)(C)C

InChI

1S/C8H14N4OS/c1-8(2,3)5-6(13)12(9)7(14-4)11-10-5/h9H2,1-4H3

InChI key

FOXFZRUHNHCZPX-UHFFFAOYSA-N

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com.

Other Notes

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany


signalword

Warning

Storage Class

11 - Combustible Solids

flash_point_f

Not applicable

flash_point_c

Not applicable

target_organs

Blood

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 2



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
802809100004022536376189
02489-5ML04061838617583
240613-100ML04061823361064
802809250004022536376196
802809010004022536376172
C68563-500ML04061833511657
C68563-1L04061833511640

Questions

Reviews

No rating value

Active Filters