Skip to Content
Merck

Skip To

74601

3,3′-Diindolylmethane

analytical standard

Synonym(s):

3,3′-Bisindolylmethane, DIM

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
100 mg
Please contact Customer Service for Availability
CZK 3,490.00

About This Item

Empirical Formula (Hill Notation):
C17H14N2
CAS Number:
Molecular Weight:
246.31
UNSPSC Code:
41116107
NACRES:
NA.24
MDL number:

CZK 3,490.00


Please contact Customer Service for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

analytical standard

Quality Segment

assay

≥97.5% (HPLC)

shelf life

limited shelf life, expiry date on the label

application(s)

food and beverages

format

neat

storage temp.

2-8°C

SMILES string

C(c1c[nH]c2ccccc12)c3c[nH]c4ccccc34

InChI

1S/C17H14N2/c1-3-7-16-14(5-1)12(10-18-16)9-13-11-19-17-8-4-2-6-15(13)17/h1-8,10-11,18-19H,9H2

InChI key

VFTRKSBEFQDZKX-UHFFFAOYSA-N

General description

3,3′-Diindolylmethane, a dimer of indole-3-carbinol, is a component of cruciferous vegetables. It is known to exhibit anticancer effects.

Application

3,3′-Diindolylmethane may be used as a model substrate to analyze the role of 3,3′-diindolylmethane in inducing apoptosis in human cancer cells.

Biochem/physiol Actions

Acid-catalyzed reaction product of a phytochemical naturally found in Brassicaceae, indole-3-carbinol. It functions as an antitumor agent. This derivative can both directly stimulate apoptosis at relatively high concentrations and sensitize TRAIL-induced apoptosis in human cancer cells. DIM induces a G1 cell cycle arrest in human breast cancer MCF-7 cells by a mechanism that includes increased expression of p21. DIM is a strong mitochondrial H+-ATPase inhibitor. The function of DIM and its derivatives as a new plant growth promoter has been studied in an eco-friendly system.

Other Notes

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
D9568BCR29776854
grade

analytical standard

grade

-

grade

certified reference material

grade

-

format

neat

format

-

format

neat

format

-

shelf life

limited shelf life, expiry date on the label

shelf life

-

shelf life

-

shelf life

-

Quality Level

100

Quality Level

100

Quality Level

-

Quality Level

100

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

-

application(s)

food and beverages

application(s)

-

application(s)

-

application(s)

-


Still not finding the right product?

Explore all of our products under 3,3′-Diindolylmethane


pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Aquatic Chronic 4 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters