Skip to Content
Merck

Skip To

73103

Dibenzyl ethoxymethylenemalonate

derivatization grade (HPLC), LiChropur, ≥98.0% (HPLC)

Synonym(s):

Bis(phenylmethyl) 2-(ethoxymethylidene)propanedioate, DBEEM, Ethoxymethylenemalonic acid dibenzyl ester

Sign In to View Organizational & Contract Pricing.

About This Item

Empirical Formula (Hill Notation):
C20H20O5
CAS Number:
Molecular Weight:
340.37
UNSPSC Code:
41121813
NACRES:
NA.22


grade

derivatization grade (HPLC)

Quality Level

assay

≥98.0% (HPLC)

form

liquid

quality

LiChropur

technique(s)

HPLC: suitable

SMILES string

O=C(OCC1=CC=CC=C1)C(C(OCC2=CC=CC=C2)=O)=COCC

Other Notes

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany


hcodes

Hazard Classifications

Aquatic Chronic 4

Storage Class

12 - Non Combustible Liquids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Riin Rebane et al.
Journal of chromatography. A, 1390, 62-70 (2015-03-12)
Derivatization is one of the most common ways for improving chromatographic separation and sensitivity for LC-ESI-MS analysis. The aim of this work was to design new derivatization reagents for LC-ESI-MS analysis of amino acids which would (1) provide good reversed



Global Trade Item Number

SKUGTIN
73103-50MG04061833415870

Questions

Reviews

No rating value

Active Filters