Skip to Content
Merck

Skip To

69965

Mossel Broth

suitable for microbiology, NutriSelect® Basic

Synonym(s):

Buffered Glucose-Brilliant Green Bile Broth, EE Broth, Enterobacteriaceae Enrichment Broth acc. to Mossel

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice
500 g
Please contact Customer Service for Availability
CZK 4,670.00

About This Item

NACRES:
NA.74
UNSPSC Code:
41171607

CZK 4,670.00


Please contact Customer Service for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


sterility

non-sterile

Quality Segment

form

powder

shelf life

limited shelf life, expiry date on the label

composition

brilliant green, 0.015 g/L , disodium hydrogen phosphate, 6.45 g/L , glucose, 5 g/L , ox-bile, 20 g/L , peptone, 10 g/L , potassium dihydrogen phosphate, 2 g/L

manufacturer/tradename

NutriSelect® Basic

technique(s)

microbiological culture: suitable

final pH

7.2±0.2 (20 °C)

application(s)

agriculture
bioburden testing
environmental
food and beverages
water monitoring, microbiology

suitability

selective for Citrobacter spp., selective for Enterococcus spp., selective for Escherichia coli, selective for Klebsiella spp., selective for Proteus spp., selective for Salmonella spp., selective for Shigella spp., selective for Yersinia spp., selective for coliforms, selective for enterobacteriaceae

Application

For the selective enrichment of all enterobacteriaceae from food and other material.

Preparation Note

Dissolve 43.5 g in 1 litre distilled water and heat at 100°C in water bath or flowing steam for 30 minutes. Cool quickly. This medium is heat sensitive. DO NOT AUTOCLAVE.

Other Notes

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.
Isolation of Serratia marcescens from clinical samples

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

This product will be discontinued in the near future; however, stock is still available. The recommended replacement product is 1.05394 - Enterobacteriaceae Enrichment Mossel broth

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
171627670444653
form

powder

form

powder

form

powder

form

powder

technique(s)

microbiological culture: suitable

technique(s)

microbiological culture: suitable

technique(s)

microbiological culture: suitable

technique(s)

microbiological culture: suitable

suitability

selective for Citrobacter spp., selective for Escherichia coli, selective for Proteus spp., selective for Shigella spp., selective for coliforms, selective for Enterococcus spp., selective for Salmonella spp., selective for enterobacteriaceae, selective for Klebsiella spp., selective for Yersinia spp.

suitability

selective for Escherichia coli, selective for coliforms

suitability

selective for Escherichia coli, selective for coliforms, selective for enterovirulent E. coli (EEC)

suitability

selective for Escherichia coli, selective for coliforms

composition

brilliant green, 0.015 g/L , glucose, 5 g/L , peptone, 10 g/L , disodium hydrogen phosphate, 6.45 g/L , potassium dihydrogen phosphate, 2 g/L , ox-bile, 20 g/L

composition

basic fuchsin, 1 g/L , dipotassium phosphate, 7 g/L , lactose, 25 g/L , peptic digest of animal tissue, 20 g/L , sodium sulfite, 2.5 g/L , yeast extract, 6 g/L

composition

bile salts No. 3, 1.5 g/L , di-potassium hydrogen phosphate, 4.0 g/L , D(+)-glucose, 2.5 g/L , novobiocin, 0.02 g/L , peptone from casein, 17.0 g/L , peptone from soymeal, 3.0 g/L , sodium chloride, 5.0 g/L

composition

bile salts, 1.5 g/L , casein digest (pancreatic), 20 g/L , dipotassium phosphate, 4 g/L , lactose, 5 g/L , potassium phosphate, 1.5 g/L , sodium chloride, 5 g/L

sterility

non-sterile

sterility

-

sterility

non-sterile

sterility

non-sterile

application(s)

agriculture
bioburden testing
environmental
food and beverages
water monitoring, microbiology

application(s)

agriculture
environmental
food and beverages
water monitoring, microbiology

application(s)

agriculture
food and beverages
microbiology
pathogen testing

application(s)

agriculture
bioburden testing
environmental
food and beverages
water monitoring, microbiology


Still not finding the right product?

Explore all of our products under Mossel Broth


hcodes

Hazard Classifications

Aquatic Chronic 3

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library


Protocols

General guidance for the detection and enumeration of Enterobacteriaceae in food according to EN-ISO 8523:1991 and EN-ISO 4832:1991, respectively.

Articles

Cronobacter spp. are frequently found on vegetables, meat, fermented bread, dairy and in baby food. Consumption of contaminated powdered infant formula milk can result in sepsis, infant meningitis and necrotizing enterocolitis.

An article concerning selective growth media for differentiation and detection of Escherichia coli and other coliforms.


G.M. Grasso et al.
Epidem. Inf., 101, 315-315 (1988)
D.A.A. Mossel et al.
The Journal of Applied Bacteriology, 26, 444-444 (1963)



Global Trade Item Number

SKUGTIN
69965-500G04061832795133

Questions

Reviews

No rating value

Active Filters