Skip to Content
Merck
All Photos(2)

Key Documents

67319

Supelco

2-Furanmethanethiol

analytical standard

Synonym(s):

2-Furfurylthiol, 2-Furylmethanethiol, Furfuryl mercaptan

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H6OS
CAS Number:
Molecular Weight:
114.17
Beilstein:
383594
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥98.0% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.531 (lit.)
n20/D 1.531-1.535

bp

155 °C (lit.)

density

1.132 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

storage temp.

2-8°C

SMILES string

SCc1ccco1

InChI

1S/C5H6OS/c7-4-5-2-1-3-6-5/h1-3,7H,4H2

InChI key

ZFFTZDQKIXPDAF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

113.0 °F - closed cup

Flash Point(C)

45 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Brian G Lake et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 41(12), 1761-1770 (2003-10-18)
The metabolism of two thiofurans, namely furfuryl mercaptan (FM) and 2-methyl-3-furanthiol (MTF), to their corresponding methyl sulphide and methyl sulphoxide derivatives has been studied in male Sprague-Dawley rat hepatocytes and liver microsomes. Rat hepatocytes converted FM to furfuryl methyl sulphoxide
Luigi Poisson et al.
Journal of agricultural and food chemistry, 57(21), 9923-9931 (2009-10-13)
The formation of several key odorants, such as 2-furfurylthiol (FFT), alkylpyrazines, and diketones, was studied upon coffee roasting. The approach involved the incorporation of potential precursors in green coffee beans by means of biomimetic in-bean and spiking experiments. Both labeled
Tuong Huynh-Ba et al.
Journal of agricultural and food chemistry, 51(12), 3629-3635 (2003-05-29)
Baker's yeast was shown to catalyze the transformation of cysteine-furfural conjugate into 2-furfurylthiol. The biotransformation's yield and kinetics were influenced by the reaction parameters such as pH, incubation mode (aerobic and anaerobic), and substrate concentration. 2-Furfurylthiol was obtained in an
Christoph Müller et al.
Journal of agricultural and food chemistry, 54(26), 10076-10085 (2006-12-21)
Recent investigations demonstrated that the reaction of odor-active thiols such as 2-furfurylthiol with thermally generated chlorogenic acid degradation products is responsible for the rapid aroma staling of coffee beverages. To get a clear understanding of the molecular mechanisms underlying this
Imre Blank et al.
Journal of agricultural and food chemistry, 50(8), 2356-2364 (2002-04-04)
The stability of the coffee flavor compound furfuryl mercaptan has been investigated in aqueous solutions under Fenton-type reaction conditions. The impact of hydrogen peroxide, iron, ascorbic acid, and ethylenediaminetetraacetic acid was studied in various combinations of reagents and temperature. Furfuryl

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service