Skip to Content
Merck

Skip To

66050

2-Methyl-2-butene

analytical standard

Synonym(s):

β-Isoamylene, Amylene

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice
5 mL
Please contact Customer Service for Availability
CZK 1,790.00
10 mL
Please contact Customer Service for Availability
CZK 3,080.00
50 mL
Please contact Customer Service for Availability
CZK 11,500.00

About This Item

Linear Formula:
CH3CH=C(CH3)2
CAS Number:
Molecular Weight:
70.13
UNSPSC Code:
41116107
NACRES:
NA.24
PubChem Substance ID:
EC Number:
208-156-3
Beilstein/REAXYS Number:
1361353
MDL number:

CZK 1,790.00


Please contact Customer Service for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

analytical standard

Quality Segment

vapor density

2.4 (vs air)

assay

≥99.0% (GC)

shelf life

limited shelf life, expiry date on the label

expl. lim.

8.7 %

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

refractive index

n20/D 1.385 (lit.), n20/D 1.387

bp

35-38 °C (lit.)

mp

−134 °C (lit.)

density

0.662 g/mL at 25 °C (lit.)

application(s)

environmental
petroleum

format

neat

SMILES string

C\C=C(\C)C

InChI

1S/C5H10/c1-4-5(2)3/h4H,1-3H3

InChI key

BKOOMYPCSUNDGP-UHFFFAOYSA-N

General description

2-Methyl-2-butene is a trisubstituted olefin which is mostly used as a precursor for peroxyacetyl nitrate. It can be prepared by the acid-catalyzed dehydration of 3-methyl-2-butanol. It has 3 alkyl groups on the double bond.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
It has been used in isolation and identification of carbonyl compounds reacting with ozone using off-line sampling method followed by liquid chromatography.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
862628.14874257486
technique(s)

HPLC: suitable, gas chromatography (GC): suitable

technique(s)

-

technique(s)

-

technique(s)

-

application(s)

environmental
petroleum

application(s)

-

application(s)

-

application(s)

-

format

neat

format

-

format

-

format

-

Quality Level

100

Quality Level

100

Quality Level

200

Quality Level

100

grade

analytical standard

grade

-

grade

-

grade

-

assay

≥99.0% (GC)

assay

≥95.0% (GC)

assay

83.0-88.0% (GC)

assay

98%


Still not finding the right product?

Explore all of our products under 2-Methyl-2-butene


signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Asp. Tox. 1 - Flam. Liq. 1 - Muta. 2 - STOT SE 3

target_organs

Central nervous system

Storage Class

3 - Flammable liquids

wgk

WGK 3

flash_point_f

-4.0 °F

flash_point_c

-20 °C

ppe

Eyeshields, Faceshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Brown, William H.
Study Guide with Student Solutions Manual for Brown/Foote/Iverson/Anslyn's Organic Chemistry,, 234-234 (2011)
Gas-phase reaction of ozone with 2-methyl-2-butene: Dicarbonyl formation from Criegee biradicals.
Grosjean, Daniel.
Environmental Science & Technology, 24.9, 1428-1432 (1990)
Tomoyasu Kadoguchi et al.
Experimental physiology, 100(3), 312-322 (2015-01-13)
What is the central question of this study? Does angiotensin II directly induce skeletal muscle abnormalities? What is the main finding and its importance? Angiotensin II induces skeletal muscle abnormalities and reduced exercise capacity. Mitochondrial dysfunction and a decreased number of oxidative



Global Trade Item Number

SKUGTIN
66050-50ML04061836958992
66050-10ML04061832734026
66050-5ML04061832734033

Questions

Reviews

No rating value

Active Filters