Skip to Content
Merck

Skip To

58381

Hamilton® syringe, 700 series, special needle

702SNR, volume 25 μL, needle size 22s ga (blunt tip), needle L 51 mm (2 in.)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
1 ea
Please contact Customer Service for Availability
CZK 1,610.00

About This Item

UNSPSC Code:
41122001
NACRES:
NB.03

CZK 1,610.00


Please contact Customer Service for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


material

glass body

needle size

22s ga (blunt tip)

feature

needle type SN (special needle)

packaging

pkg of 1 ea

manufacturer/tradename

Hamilton 80465

needle L

51 mm (2 in.)

volume

25 μL

compatibility

for use with Rheodyne, for use with SSI, for use with Valco VISF-2

Other Notes

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Hamilton is a registered trademark of Hamilton Co.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
207212071420715
material

glass body

material

glass body

material

glass body

material

glass body

packaging

pkg of 1 ea

packaging

pkg of 1 ea

packaging

pkg of 1 ea

packaging

pkg of 1 ea

volume

25 μL

volume

2.5 mL

volume

0.5 mL

volume

50 μL

needle L

51 mm (2 in.)

needle L

, n/a

needle L

, n/a

needle L

, n/a

needle size

22s ga (blunt tip)

needle size

(not included)

needle size

(not included)

needle size

(not included)

manufacturer/tradename

Hamilton 80465

manufacturer/tradename

Hamilton 81401

manufacturer/tradename

Hamilton 81201

manufacturer/tradename

Hamilton 80901


Still not finding the right product?




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters