Sign In to View Organizational & Contract Pricing.
About This Item
Empirical Formula (Hill Notation):
C15D3H14Cl2N3O2
Molecular Weight:
345.24
UNSPSC Code:
41116107
NACRES:
NA.24
PubChem Substance ID:
grade
analytical standard
Quality Segment
product line
PESTANAL®
assay
≥94.0% (HPLC)
shelf life
limited shelf life, expiry date on the label
application(s)
agriculture
format
neat
storage temp.
2-8°C
General description
Propiconazole-(phenyl-d3) is an isotope labeled analog of the triazole fungicide propiconazole, wherein C-3, C-5 and C-6 protons of the dichlorophenyl moiety are replaced by deuterium.
Application
Isotope-labeled compounds are increasingly used in isotope dilution mass spectrometry (IDMS) for the quantitative analysis of pesticides. The major advantage of using this technique is that the isotope-labeled compounds have nearly the same physical properties as their non-labeled counterpart analogs, and thus show identical behavior during the workup and sample preparation process. This helps in overcoming the problems of matrix effects generally encountered in the usual LC-MS/GC-MS analysis, potentially resulting in biased results. By spiking the sample before workup with its isotope labeled analog, the loss of analyte that leads to matrix effects can be determined and compensated.
Legal Information
PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany
signalword
Danger
hcodes
Hazard Classifications
Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Repr. 1B - Skin Sens. 1
Storage Class
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Regulatory Listings
Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.
2140327-43-9
CAS No.
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Residues and dissipation kinetics of triazole fungicides difenoconazole and propiconazole in wheat and soil in Chinese fields
Zhang Z, et al.
Food Chemistry, 168, 396-403 (2015)
Global Trade Item Number
| SKU | GTIN |
|---|---|
| 55833-5MG | 04061832583099 |


