Skip to Content
Merck
All Photos(1)

Documents

46770

Supelco

Sulfacetamide

VETRANAL®, analytical standard

Synonym(s):

N-(4-Aminobenzenesulfonyl)acetamide, N-(4-Aminophenylsulfonyl)acetamide, N1-Acetylsulfanilamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H10N2O3S
CAS Number:
Molecular Weight:
214.24
Beilstein:
981718
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

VETRANAL®

Assay

≥98.0%

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

clinical testing

format

neat

SMILES string

CC(=O)NS(=O)(=O)c1ccc(N)cc1

InChI

1S/C8H10N2O3S/c1-6(11)10-14(12,13)8-4-2-7(9)3-5-8/h2-5H,9H2,1H3,(H,10,11)

InChI key

SKIVFJLNDNKQPD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Chemical structure: sulfonamide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

Demet Sensoy et al.
European journal of pharmaceutics and biopharmaceutics : official journal of Arbeitsgemeinschaft fur Pharmazeutische Verfahrenstechnik e.V, 72(3), 487-495 (2009-02-19)
The aim of this study was to prepare bioadhesive sulfacetamide sodium (SA) microspheres to increase their residence time on the ocular surface and to enhance their treatment efficacy on ocular keratitis. Microspheres were fabricated by spray drying method using mixture
E Kremer et al.
Journal of inorganic biochemistry, 100(7), 1167-1175 (2006-04-06)
The synthesis, characterization and comparative biological study of a series of antibacterial copper complexes with heterocyclic sulfonamides were reported. Two kinds of complexes were obtained with the stoichiometries [Cu(L)2] . H2O and [Cu(L)2(H2O)4] . nH2O. They were characterized by infrared
Rade Injac et al.
Food additives & contaminants. Part A, Chemistry, analysis, control, exposure & risk assessment, 25(4), 424-431 (2008-03-19)
A separation technique for zinc bacitracin, polymyxin B, oxytetracycline and sulfacetamide in animal feedstuffs by micellar electrokinetic capillary chromatography (MEKC) was developed. The running buffer was 20 mmol l(-1) borate, 20 mmol l(-1) phosphate, pH 8.4, containing 20 mmol l(-1)
Meghan I Dubina et al.
Archives of dermatology, 145(9), 1027-1029 (2009-09-23)
A recent study demonstrated evidence of a yellow-orange discoloration of the skin and hair when topical dapsone gel was combined with benzoyl peroxide. This phenomenon had previously been observed by one of us (A.B.F.) when sulfasalazine was combined with benzoyl
Fatmah A M Al-Omary et al.
Bioorganic & medicinal chemistry, 18(8), 2849-2863 (2010-03-31)
A new series of 2,6-substituted-quinazolin-4-ones was designed, synthesized, and evaluated for their in vitro DHFR inhibition, antimicrobial, and antitumor activities. Compounds 22, 33-37, 39-43, and 45 proved to be active DHFR inhibitors with IC(50) range of 0.4-1.0microM. Compound 18 showed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service