Skip to Content
Merck

Skip To

43311

2-(Methacryloyloxy)ethyl acetoacetate

Arxada quality, ≥95.0% (GC), contains BHT as stabilizer

Synonym(s):

Lonzamon AAEMA, (2-Acetoacetoxy)ethyl methacrylate, Ethylene glycol monoacetoacetate monomethacrylate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice
1 kg
Please contact Customer Service for Availability
CZK 1,530.00
10 kg
Please contact Customer Service for Availability
CZK 9,900.00
25 kg
Please contact Customer Service for Availability
CZK 24,200.00

About This Item

Linear Formula:
CH3COCH2CO2CH2CH2O2CC(CH3)=CH2
CAS Number:
Molecular Weight:
214.22
NACRES:
NA.22
UNSPSC Code:
12352100
Beilstein/REAXYS Number:
2099809
MDL number:

CZK 1,530.00


Please contact Customer Service for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥95.0% (GC)

form

liquid

quality

Arxada quality

contains

BHT as stabilizer

manufacturer/tradename

Arxada AG

impurities

≤0.10% water, ≤2.5% 2-hydroxyethyl methacrylate (GC)

refractive index

n20/D 1.456 (lit.)

bp

100 °C/0.8 mmHg (lit.)

acid value

3.0‑8.0 KOH/g

density

1.122 g/mL at 25 °C (lit.)

SMILES string

CC(=O)CC(=O)OCCOC(=O)C(C)=C

InChI

1S/C10H14O5/c1-7(2)10(13)15-5-4-14-9(12)6-8(3)11/h1,4-6H2,2-3H3

InChI key

IBDVWXAVKPRHCU-UHFFFAOYSA-N


Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
8.188475374038.00857
quality

Arxada quality

quality

-

quality

-

quality

-

acid value

3.0‑8.0 KOH/g

acid value

-

acid value

-

acid value

-

form

liquid

form

liquid

form

-

form

liquid

impurities

≤0.10% water, ≤2.5% 2-hydroxyethyl methacrylate (GC)

impurities

-

impurities

-

impurities

-

contains

BHT as stabilizer

contains

-

contains

BHT as stabilizer

contains

-

Quality Level

100

Quality Level

200

Quality Level

200

Quality Level

200


Still not finding the right product?


Storage Class

10 - Combustible liquids

wgk

WGK 1

flash_point_f

231.8 °F - closed cup

flash_point_c

111 °C - closed cup



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
359300-10G04061831813210
359300-50G04061832564616
43311-10KG04065265455275
43311-25KG04065265455299
43311-1KG04065265455282

Questions

Reviews

No rating value

Active Filters