Skip to Content
Merck
All Photos(1)

Key Documents

36504

Supelco

Fenazaflor

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H7Cl2F3N2O2
CAS Number:
Molecular Weight:
375.13
Beilstein:
964952
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

storage temp.

2-8°C

SMILES string

FC(F)(F)c1nc2cc(Cl)c(Cl)cc2n1C(=O)Oc3ccccc3

InChI

1S/C15H7Cl2F3N2O2/c16-9-6-11-12(7-10(9)17)22(13(21-11)15(18,19)20)14(23)24-8-4-2-1-3-5-8/h1-7H

InChI key

ISVQSVPUDBVFFU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Fenazaflor is a proacaricide,[1] often employed in apple orchards to protect the crop against a variety of fungal diseases and insect pests.[2]

Application

Fenazaflor may be used as an analytical reference standard for the determination of the analyte in fruit juice and serum samples[3][4] using various chromatographic techniques.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

>212.0 °F

Flash Point(C)

> 100 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

[Study of the effect of phenophtorazole on the calcium ion concentration in rat thymocyte cytoplasm and on changes of this parameter, induced by concanavalin A].
E I Astashkin et al.
Doklady Akademii nauk SSSR, 317(2), 499-502 (1991-01-01)
Molecular Mechanisms of Resistance to Agrochemicals, 13 (2013)
[Effect of inhibitors of lipoxygenase oxidation of arachidonic acid and phenofluorazole on rat thymocyte cytoplasmic pH and on the change of this parameter induced by concanavalin A].
E N Astashkin et al.
Doklady Akademii nauk SSSR, 325(4), 869-873 (1992-01-01)
Chemical resistance in populations of Tetranychus urticae (Koch)(Acarina: Tetranychidae) from apple orchards in New South Wales, Australia.
Unwin B.
Austral Entomology, 12(1), 59-67 (1973)
Rapid simultaneous determination for organophosphorus pesticides in human serum by LC-MS.
Inoue S, et al.
Journal of Pharmaceutical and Biomedical Analysis, 44(1), 258-264 (2007)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service