Skip to Content
Merck

Skip To

36136

Isomethiozin

PESTANAL®, analytical standard

Sign In to View Organizational & Contract Pricing.

About This Item

Empirical Formula (Hill Notation):
C12H20N4OS
CAS Number:
Molecular Weight:
268.38
NACRES:
NA.24
UNSPSC Code:
41116107
EC Number:
260-532-6


grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

CSC1=NN=C(C(=O)N1\N=C/C(C)C)C(C)(C)C

InChI

1S/C12H20N4OS/c1-8(2)7-13-16-10(17)9(12(3,4)5)14-15-11(16)18-6/h7-8H,1-6H3/b13-7+

InChI key

MZTLOILRKLUURT-NTUHNPAUSA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Isomethiozin may be used as an analytical reference standard for the determination of the analyte in animal muscles and tissues, surface water and fruits and vegetables by various chromatographic techniques.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany


Storage Class

13 - Non Combustible Solids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Screening and quantification of 304 pesticides and related organic pollutants in surface water using dispersive liquid--liquid microextraction coupled with gas chromatography-mass spectrometry.
Chen B, et al.
Analytical Methods : Advancing Methods and Applications, 6(6), 1743-1752 (2014)
Determination of isomethiozin by differential-pulse polarography.
J F Arranz Valentín et al.
The Analyst, 113(4), 629-632 (1988-04-01)
Analysis method study on 839 pesticide and chemical contaminant multiresidues in animal muscles by gel permeation chromatography cleanup, GC/MS, and LC/MS/MS.
Pang GF, et al.
Journal of AOAC (Association of Official Analytical Chemists) International, 92(3), S1-S72 (2009)



Global Trade Item Number

SKUGTIN
36136-100MG04061831824155

Questions

Reviews

No rating value

Active Filters