Skip to Content
Merck

Skip To

32771

Aflatoxin B2-13C17 solution

~0.5 μg/mL in acetonitrile, analytical standard

Synonym(s):

Fully 13C-labelled Aflatoxin B2

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice
1 mL
Please contact Customer Service for Availability
CZK 77,800.00

About This Item

Empirical Formula (Hill Notation):
13C17H14O6
CAS Number:
Molecular Weight:
331.16
NACRES:
NA.24
PubChem Substance ID:
UNSPSC Code:
85151701
EC Number:
200-835-2
MDL number:

CZK 77,800.00


Please contact Customer Service for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

analytical standard

Quality Level

shelf life

limited shelf life, expiry date on the label

concentration

~0.5 μg/mL in acetonitrile

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

single component solution

mass shift

M+17

storage temp.

−20°C

SMILES string

[13CH3]O[13c]1[13cH][13c]2O[13C@H]3O[13CH2][13CH2][13C@H]3[13c]2[13c]4O[13C](=O)[13C]5=[13C]([13CH2][13CH2][13C]5=O)[13c]14

InChI

1S/C17H14O6/c1-20-10-6-11-14(8-4-5-21-17(8)22-11)15-13(10)7-2-3-9(18)12(7)16(19)23-15/h6,8,17H,2-5H2,1H3/t8-,17+/m0/s1/i1+1,2+1,3+1,4+1,5+1,6+1,7+1,8+1,9+1,10+1,11+1,12+1,13+1,14+1,15+1,16+1,17+1

InChI key

WWSYXEZEXMQWHT-DENWKYDXSA-N

General description

Aflatoxin B2-13C17 is the 13C isotope-labeled analog of the mycotoxin, aflatoxin.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Aflatoxin B2-13C17 may be used as an analytical standard for the determination of aflatoxin B2 in food grade gums[1] as well as milk, milk-based infant formula, and feed samples by stable isotope dilution and liquid chromatography combined with tandem mass spectrometry (LC-MS/MS).

Analysis Note

purity : ≥97.0% (HPLC)

Disclaimer

Supply conditions do not apply to the regions and states of Brazil: North, Northeast, Mato Grosso do Sul, Mato Grosso, and Rio Grande do Sul.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
327773276432772
technique(s)

HPLC: suitable, gas chromatography (GC): suitable

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

grade

analytical standard

grade

analytical standard

grade

analytical standard

grade

analytical standard

format

single component solution

format

single component solution

format

single component solution

format

single component solution

application(s)

cleaning products
cosmetics
food and beverages
personal care

application(s)

cleaning products
cosmetics
food and beverages
personal care

application(s)

cleaning products
cosmetics
food and beverages
personal care

application(s)

cleaning products
cosmetics
food and beverages
personal care

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

concentration

~0.5 μg/mL in acetonitrile

concentration

~0.5 μg/mL in acetonitrile

concentration

~0.5 μg/mL in acetonitrile

concentration

~0.5 μg/mL in acetonitrile


pictograms

FlameExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class

3 - Flammable liquids

wgk

WGK 2

flash_point_f

35.6 °F - closed cup

flash_point_c

2.0 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Interlaboratory Validation of a Stable Isotope Dilution and Liquid Chromatography Tandem Mass Spectrometry Method for the Determination of Aflatoxins in Milk, Milk-Based Infant Formula, and Feed.
Zhang K, et al.
Journal of AOAC (Association of Official Analytical Chemists) International, 101(3), 677-685 (2018)
Kai Zhang et al.
Journal of AOAC International, 97(3), 889-895 (2014-07-24)
Stable isotope dilution with LC/MSIMS was used to determine the following 11 mycotoxins in food grade gums: aflatoxins B1, B2, G1, and G2; deoxynivalenol; fumonisins B1, B2, and B3; ochratoxin A; T-2 toxin; and zearalenone. Samples were fortified with 11



Global Trade Item Number

SKUGTIN
32771-1ML04061826717066

Questions

Reviews

No rating value

Active Filters