Skip to Content
Merck
All Photos(3)

Key Documents

294098

Sigma-Aldrich

Benzonitrile

anhydrous, ≥99%

Synonym(s):

Phenyl cyanide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CN
CAS Number:
Molecular Weight:
103.12
Beilstein:
506893
EC Number:
MDL number:
UNSPSC Code:
12352002
eCl@ss:
39031505
PubChem Substance ID:
NACRES:
NA.21
grade:
anhydrous
Assay:
≥99%
bp:
191 °C (lit.)

grade

anhydrous

Quality Level

Assay

≥99%

form

liquid

expl. lim.

0.34-6.3 %

impurities

<0.003% water
<0.005% water (100 mL pkg)

evapn. residue

<0.0003%

refractive index

n20/D 1.528 (lit.)

bp

191 °C (lit.)

mp

−13 °C (lit.)

SMILES string

N#Cc1ccccc1

InChI

1S/C7H5N/c8-6-7-4-2-1-3-5-7/h1-5H

InChI key

JFDZBHWFFUWGJE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Benzonitrile may be used in the synthesis of organic building blocks such as 2-cyclopentylacetophenone, 4-carbomethoxy-5-methoxy-2-phenyl-1,3-oxazole and 1-phenyl-3,4-dihydro-6,7-methylenedioxyisoquinoline. It may also be used as a solvent in the synthesis of bis(trifluoromethyl)diazomethane.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 1

Flash Point(F)

158.0 °F - closed cup

Flash Point(C)

70 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bis(trifluoromethyl)diazomethane
Middleton WJ and Gale DM
Organic Syntheses, 50, 6-6 (1970)
Rhodium-catalyzed heterocycloaddition of a diazomalonate and a nitrile: 4-carbomethoxy-5-methoxy-2-phenyl-1,3-oxazole
Tullis JS and Helquist P
Organic Syntheses, 74, 229-229 (1997)
Generation and cyclization of 5-hexenyllithium: 2-cyclopentylacetophenone
Bailey WF, et al
Organic Syntheses, 81, 121-121 (2005)
Selective halogen-lithium exchange reactions of 2-(2'-halophenyl)ethyl halides: synthesis of 4,5-methylenedioxybenzocyclobutene and 1-phenyl-3,4-dihydro-6,7-methylenedioxyisoquinoline
Jakiela DJ, et al
Organic Syntheses, 62, 74-74 (1984)
Kenji Ueura et al.
Organic letters, 7(11), 2229-2231 (2005-05-20)
[reaction: see text]. The Suzuki-Miyaura-type cross-coupling of arylboron compounds with aryl halides proceeds efficiently in the presence of a rhodium-based catalyst system to produce the corresponding biaryls. Furthermore, it has unexpectedly been observed that the treatment with benzonitrile under similar

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service