Key Documents
20703
Hamilton® syringe, luer tip
725LT, volume 250 μL, needle size (not included)
Select a Size
1 EA
CZK 2,110.00
CZK 2,110.00
Please contact Customer Service for Availability
Request a Bulk Order
Select a Size
Change View
1 EA
CZK 2,110.00
About This Item
UNSPSC Code:
41122001
CZK 2,110.00
Please contact Customer Service for Availability
Request a Bulk Order
material
glass body
needle size
(not included)
feature
without needle Luer tip
packaging
pkg of 1 ea
manufacturer/tradename
Hamilton 80701
needle L
, n/a
volume
250 μL
compatibility
for use with Hamilton 725LT (Luer tip)
General description
Hamilton® syringe is available in gas-tight type that minimizes the gas exchange in GC analysis. It gives accurate volume delivery. The plunger is mostly unsurpassed.[1] The tip is threaded in a luer lock syringe allowing the needle to be tightly screwed to the tip.[2]
Legal Information
Hamilton is a registered trademark of Hamilton Co.
related product
Product No.
Description
Pricing
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
Jeffrey S. Guy
Pharmacology for the Prehospital Professional, 54-54 (2009)
Don P. Wolf
In Vitro Fertilization and Embryo Transfer: A Manual of Basic Techniques, 192 -192 (2012)