Skip to Content
Merck

Skip To

09773

Fmoc-D-Bpa-OH

≥98.0% (HPLC)

Synonym(s):

Fmoc-4-benzoyl-D-phenylalanine

Sign In to View Organizational & Contract Pricing.

About This Item

Empirical Formula (Hill Notation):
C31H25NO5
CAS Number:
Molecular Weight:
491.53
NACRES:
NA.26
PubChem Substance ID:
UNSPSC Code:
12352200
MDL number:


Quality Segment

assay

≥98.0% (HPLC)

form

powder

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

color

white to faint beige

application(s)

peptide synthesis

functional group

Fmoc

storage temp.

2-8°C

SMILES string

OC(=O)[C@@H](Cc1ccc(cc1)C(=O)c2ccccc2)NC(=O)OCC3c4ccccc4-c5ccccc35

InChI

1S/C31H25NO5/c33-29(21-8-2-1-3-9-21)22-16-14-20(15-17-22)18-28(30(34)35)32-31(36)37-19-27-25-12-6-4-10-23(25)24-11-5-7-13-26(24)27/h1-17,27-28H,18-19H2,(H,32,36)(H,34,35)/t28-/m1/s1

InChI key

SYOBJKCXNRQOGA-MUUNZHRXSA-N



Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
09773-1G04061826100387

Questions

Reviews

No rating value

Active Filters