Skip to Content
Merck

Skip To

08137

2-(6-Methoxynaphthalen-2-yl)acetic acid

pharmaceutical impurity standard

Synonym(s):

Naproxen impurity I, 6-Methoxy-2-naphthaleneacetic acid, 6-Methoxy-2-naphthylacetic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice
50 mg
Please contact Customer Service for Availability
CZK 13,500.00

About This Item

Empirical Formula (Hill Notation):
C13H12O3
CAS Number:
Molecular Weight:
216.23
PubChem Substance ID:
UNSPSC Code:
12352106
NACRES:
NA.24
EC Number:
245-967-1
MDL number:

CZK 13,500.00


Please contact Customer Service for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

API family

nabumetone

assay

≥98.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

impurities

≤0.5% water

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

InChI

1S/C13H12O3/c1-16-12-5-4-10-6-9(7-13(14)15)2-3-11(10)8-12/h2-6,8H,7H2,1H3,(H,14,15)

InChI key

PHJFLPMVEFKEPL-UHFFFAOYSA-N


Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
PHR2522284785N8280
format

neat

format

-

format

-

format

-

impurities

≤0.5% water

impurities

-

impurities

-

impurities

-

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

-

storage temp.

-

Quality Level

100

Quality Level

300

Quality Level

200

Quality Level

200

API family

nabumetone

API family

naproxen

API family

-

API family

-

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical small molecule

application(s)

-

application(s)

pharmaceutical (small molecule)


pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
A72405-10G04061833383896
A72405-50G04061832281469
08137-50MG04061834243151

Questions

Reviews

No rating value

Active Filters