Skip to Content
Merck

Skip To

07506

(S)-Naproxen ethyl ester

pharmaceutical impurity standard

Synonym(s):

Naproxen impurity F (PhEur), (2S)-2-(6-Methoxy-2-naphthyl)propionic acid ethyl ester, Ethyl (αS)-6-methoxy-α-methyl-2-naphthaleneacetate, Ethyl (2S)-2-(6-methoxy-2-naphthyl)propionate

Sign In to View Organizational & Contract Pricing.

About This Item

Empirical Formula (Hill Notation):
C16H18O3
CAS Number:
Molecular Weight:
258.31
NACRES:
NA.24
PubChem Substance ID:
UNSPSC Code:
12352200
MDL number:


Quality Segment

API family

naproxen

assay

≥98.0% (HPLC)

optical activity

[α]/D 48±4°, c = 0.1 in chloroform

shelf life

limited shelf life, expiry date on the label

impurities

≤0.5% water

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

C[C@H](C(OCC)=O)C1=CC2=CC=C(OC)C=C2C=C1

InChI

1S/C16H18O3/c1-4-19-16(17)11(2)12-5-6-14-10-15(18-3)8-7-13(14)9-12/h5-11H,4H2,1-3H3/t11-/m0/s1

InChI key

URNAYRDBUUZOIU-NSHDSACASA-N



Storage Class

13 - Non Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters