Select a Size
| Size/SKU | Availability | Price |
|---|---|---|
100 mL | Please contact Customer Service for Availability | CZK 2,360.00 |
About This Item
Quality Segment
product line
TraceCERT®
grade
certified reference material, TraceCERT®
concentration
1000 mg/L malate in water (nominal concentration)
technique(s)
ion chromatography: suitable
application(s)
cleaning products
cosmetics
environmental
food and beverages
personal care
format
single component solution
General description
Certified content incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com.
Analysis Note
Other Notes
Legal Information
1 of 1
This Item | |||
|---|---|---|---|
| technique(s) ion chromatography: suitable | technique(s) ion chromatography: suitable | technique(s) ion chromatography: suitable | technique(s) ion chromatography: suitable |
| application(s) cleaning products | application(s) environmental | application(s) environmental | application(s) PFAS testing |
| format single component solution | format single component solution | format single component solution | format single component solution |
| grade certified reference material, TraceCERT® | grade certified reference material, TraceCERT® | grade certified reference material, TraceCERT® | grade certified reference material, TraceCERT® |
| Quality Level 300 | Quality Level 300 | Quality Level 300 | Quality Level 300 |
| concentration 1000 mg/L malate in water (nominal concentration) | concentration 1000 mg/L chlorate in water (nominal concentration) | concentration 1000 mg/L Li+ in nitric acid (nominal concentration) | concentration 1000 mg/L fluoride in water (nominal concentration) |
Storage Class
12 - Non Combustible Liquids
wgk
nwg
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.



