Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Pack Size | SKU | Availability | Price |
|---|---|---|---|
| 50 mg | Please contact Customer Service for Availability | CZK 10,900.00 |
About This Item
Empirical Formula (Hill Notation):
C13H9Cl2NO
CAS Number:
Molecular Weight:
266.12
NACRES:
NA.24
UNSPSC Code:
41116107
Beilstein/REAXYS Number:
2121741
MDL number:
Quality Segment
assay
≥98.0% (HPLC)
shelf life
limited shelf life, expiry date on the label
impurities
≤0.5% water
application(s)
clinical testing
format
neat
SMILES string
ClC1=C(NC2=CC=CC=C2C=O)C(Cl)=CC=C1
InChI
1S/C13H9Cl2NO/c14-10-5-3-6-11(15)13(10)16-12-7-2-1-4-9(12)8-17/h1-8,16H
InChI key
MILOIOYXRHLAAH-UHFFFAOYSA-N
1 of 1
This Item | |||
|---|---|---|---|
| format neat | format - | format - | format - |
| impurities ≤0.5% water | impurities - | impurities - | impurities - |
| shelf life limited shelf life, expiry date on the label | shelf life - | shelf life - | shelf life - |
| Quality Level 100 | Quality Level 300 | Quality Level 300 | Quality Level 300 |
| assay ≥98.0% (HPLC) | assay - | assay - | assay - |
| application(s) clinical testing | application(s) pharmaceutical small molecule | application(s) pharmaceutical small molecule | application(s) pharmaceutical small molecule |
signalword
Warning
hcodes
pcodes
Hazard Classifications
Acute Tox. 4 Oral - Aquatic Chronic 4
Storage Class
11 - Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Global Trade Item Number
| SKU | GTIN |
|---|---|
| 05136-50MG | 04061834079187 |
![2-[(2,6-Dichlorophenyl)amino]benzaldehyde pharmaceutical impurity standard](/deepweb/assets/sigmaaldrich/product/structures/293/848/d04fb3ac-d732-41b5-99bd-b6f7fd052a25/640/d04fb3ac-d732-41b5-99bd-b6f7fd052a25.png)


