Skip to Content
Merck

Skip To

00929

cis-4-Hexen-1-ol

≥95.0% (GC)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C6H12O
CAS Number:
Molecular Weight:
100.16
EC Number:
213-187-0
UNSPSC Code:
12352100
PubChem Substance ID:
Beilstein/REAXYS Number:
1720136
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

≥95.0% (GC)

refractive index

n20/D 1.443

bp

80 °C/30 mmHg (lit.)

density

0.857 g/mL at 20 °C (lit.)

SMILES string

C\C=C/CCCO

InChI

1S/C6H12O/c1-2-3-4-5-6-7/h2-3,7H,4-6H2,1H3/b3-2-

InChI key

VTIODUHBZHNXFP-IHWYPQMZSA-N



pictograms

Flame

signalword

Warning

hcodes

Hazard Classifications

Flam. Liq. 3

Storage Class

3 - Flammable liquids

wgk

WGK 3

flash_point_f

98.6 °F - closed cup

flash_point_c

37 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
215163-100G04061831828863
215163-25G04061838773487