Skip to Content
Merck
All Photos(1)

Key Documents

00670590

Thymol

primary reference standard

Synonym(s):

2-Isopropyl-5-methylphenol, 5-Methyl-2-(1-methylethyl)phenol, 5-Methyl-2-isopropylphenol, IPMP

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
2-[(CH3)2CH]C6H3-5-(CH3)OH
CAS Number:
Molecular Weight:
150.22
Beilstein:
1907135
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

primary reference standard

vapor pressure

1 mmHg ( 64 °C)

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

HWI

bp

232 °C (lit.)

mp

48-51 °C (lit.)

density

0.965 g/mL at 25 °C (lit.)

SMILES string

CC(C)c1ccc(C)cc1O

InChI

1S/C10H14O/c1-7(2)9-5-4-8(3)6-10(9)11/h4-7,11H,1-3H3

InChI key

MGSRCZKZVOBKFT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Produced and qualified by HWI pharma services GmbH.
Exact content by quantitative NMR can be found on the certificate.

Application

Reference Standard in the analysis of herbal medicinal products

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

240.8 °F - closed cup

Flash Point(C)

116 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Hiroko Ohwada et al.
Research in developmental disabilities, 34(1), 650-655 (2012-11-06)
As the life expectancy of people with intellectual disability (ID) increases, it is becoming necessary to understand factors affecting survival. However, predictors that are typically assessed among healthy people have not been examined. Predictors of all-cause mortality, including blood, urine
María C Elissondo et al.
Acta tropica, 125(3), 251-257 (2012-12-25)
Scolicidal solutions remain indispensable in the treatment of hydatid cyst disease. Properties of an ideal solution would be inexpensiveness and the promotion of a rapid and complete scolicidal effect with an absence of local and systemic side effects. From this
Bhavini Shah et al.
Journal of food protection, 76(3), 440-447 (2013-03-07)
Food safety is a continuing challenge for the food industry due to sporadic illness outbreaks caused by foodborne pathogens. Plant essential oils have been studied extensively as natural antimicrobials to control foodborne pathogens. However, their hydrophobic nature makes application in
H Hashemipour et al.
Poultry science, 92(8), 2059-2069 (2013-07-23)
This trial was conducted to evaluate the effects of dietary supplementation of phytogenic product containing an equal mixture of thymol and carvacrol at 4 levels (0, 60, 100, and 200 mg/kg of diet) on performance, antioxidant enzyme activities, fatty acid
S Gentile et al.
Biochemical pharmacology, 34(14), 2439-2444 (1985-07-15)
The binding of the colored cholephylic anions tetrabromosulfonphthalein (BSP), di-bromosulfonphthalein (DBSP), indocyanine green (ICG) and thymol blue (ThB) to a number of protein preparations including bovine serum albumin, human serum, rat hepatic cytosol and purified rat liver bilitranslocase has been

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service