Skip to Content
Merck
All Photos(1)

Key Documents

24-5110

Sigma-Aldrich

Potassium periodate

JIS special grade, 99.5-100.3%

Synonym(s):

Periodic acid potassium salt, Potassium (meta)periodate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
KIO4
CAS Number:
Molecular Weight:
230.00
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
Assay:
99.5-100.3%
grade:
JIS special grade
form:
powder

grade

JIS special grade

Assay

99.5-100.3%

form

powder

reaction suitability

reagent type: oxidant

availability

available only in Japan

pH

4.5-5.5 (25 °C, 2.3 g/L)

mp

582 °C (lit.)

SMILES string

[K+].[O-]I(=O)(=O)=O

InChI

1S/HIO4.K/c2-1(3,4)5;/h(H,2,3,4,5);/q;+1/p-1

InChI key

FJVZDOGVDJCCCR-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Eye Dam. 1 - Ox. Sol. 1 - Skin Corr. 1C - STOT RE 1

Target Organs

Thyroid

Storage Class Code

5.1A - Strongly oxidizing hazardous materials

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jianxiu Du et al.
Luminescence : the journal of biological and chemical luminescence, 25(1), 76-80 (2009-07-25)
A simple and sensitive chemiluminescence (CL) method combined with flow injection technique was developed for the determination of naproxen. It was based upon the weak CL signal arising from the reaction of KIO(4) with H(2)O(2) being significantly increased by naproxen
Ming Li et al.
Journal of fluorescence, 14(5), 597-601 (2004-12-25)
A method to determine glucose using an optical sensor prepared by entrapping glucose oxidase into silica sol-gel column has been developed. The silica sol-gel film was coated on alumina substrate. The optical sensor is based on the chemiluminescence intensity from
Yan-Ming Liu et al.
Electrophoresis, 28(12), 1937-1941 (2007-05-08)
A rapid and sensitive method to detect three catecholamines, isoprenaline, epinephrine, and dopamine, by CE coupled with direct luminol-potassium periodate chemiluminescence (CL) detection is described. The conditions for CE separation and CL reaction were systematically optimized. Under the optimum conditions
K Fukuyama et al.
The Journal of biological chemistry, 270(37), 21884-21892 (1995-09-15)
The structures of the cyanide and triiodide complexes of Arthromyces ramosus peroxidase (ARP) at different pH values were investigated by x-ray crystallography in order to examine the behavior of the invariant residues of arginine (Arg-52) and distal histidine (His-56) during
Tsung-Yuan Huang et al.
Luminescence : the journal of biological and chemical luminescence, 26(2), 118-124 (2010-02-16)
A novel phenomenon of dual chemiluminescence (CL) was observed for the KIO4-luminol-Mn(2+) system in strong alkaline solutions using the stopped-flow technique. Scavenging study of the reactive oxygen species (ROS) suggested that the two CL peaks originated from different CL pathways

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service