XX63LK230
Lab Incubator
Dual Chamber, AC/DC input 230 V, 50 Hz
About This Item
Recommended Products
Quality Level
technique(s)
analytical sample preparation: suitable
chamber size
2
input
sample type beverage(s)
sample type sport drink
sample type wine
sample type cider
sample type cosmetics
sample type soft drink
sample type beer
sample type bottled water
sample type food(s)
application(s)
cosmetics
environmental
food and beverages
industrial qc (product testing)
industrial qc (microbial monitoring)
industrial qc
water monitoring
General description
Application
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service