Key Documents
TQNXA4RP2
Millipore
Steritest® Equinox Advanced Requalification Protocol
A4 format
Select a Size
1 UNIT
CZK 19,300.00
CZK 19,300.00
Estimated to ship onMarch 12, 2025Details
Request more information
Select a Size
Change View
1 UNIT
CZK 19,300.00
About This Item
UNSPSC Code:
41116500
NACRES:
NB.79
CZK 19,300.00
Estimated to ship onMarch 12, 2025Details
Request more information
compatibility
for use with Steritest® Equinox pump.
General description
Steritest® Equinox Advanced Requalification protocol is designed as a ready to use guide to perform the requalification of the Steritest® Equinox pump. The requalification protocol can be used to re-certify consistent performance of the pumps upon encountering any change or at regular intervals. On top of some IQ and OQ tests it also covers the periodic review of operator training, SOPs, and OOS results.
Application
Steritest® Equinox Advanced Requalification protocol is used for the re-qualification of Steritest® Equinox pump which is used to perform sterility testing for pharmaceutical products.
Legal Information
STERITEST is a registered trademark of Merck KGaA, Darmstadt, Germany