Skip to Content
Merck
All Photos(1)

Key Documents

SYMBWFS01

Millipore

Steritest® Symbio waste overfilling sensor kit

For solid container, for use with Steritest® Symbio pump

Synonym(s):

Filling monitoring aid

Sign Into View Organizational & Contract Pricing

Select a Size

1 UNIT
CZK 14,900.00

CZK 14,900.00


Estimated to ship onApril 22, 2025Details


Request more information

Select a Size

Change View
1 UNIT
CZK 14,900.00

About This Item

UNSPSC Code:
23151818
NACRES:
NB.79

CZK 14,900.00


Estimated to ship onApril 22, 2025Details


Request more information

material

PBT housing (reinforced glass fibers)
polycarbonate ( hood, reinforced glass fibers)
thermoplastic polyurethane (TPU) (Buttons)

Agency

EP (2.6.1)
JP (4.06)
USP (71)

packaging

pkg of 1 kit

manufacturer/tradename

Steritest®

input

pharmaceutical(s)

application(s)

pharmaceutical
sterility testing

compatibility

for use with Steritest® Symbio FLEX Pump Kit, 2 media (SYMBFLE01)
for use with Steritest® Symbio ISL Pump Kit, 2 media (SYMBISL01)
for use with Steritest® Symbio LFH Pump Kit (SYMBLFH01)
for use with Steritest® Symbio pump

shipped in

ambient

General description

Steritest® Symbio waste overfilling sensor kit for solid container is an accessory of Steritest® Symbio Pumps. The kit prevents liquid spillage and test interruption by warning the user via both an audible signal and a visual alert on the Steritest® Symbio Pump screen when the waste container is almost full. Test in progress can be finished before the waste container is emptied or replaced.

Application

Steritest® Symbio waste overfilling sensor kit for solid container is used to prevent the liquid waste container from overflowing and is especially helpful when the waste container is placed out of the operator′s view.

Legal Information

STERITEST is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service