SM-2002
Trypsin-EDTA, In Hank′s Balanced Salt Solution, 0.05% Trypsin and 0.53mM EDTA, without Ca 2+ and Mg 2+
The Trypsin-EDTA, In Hank′s Balanced Salt Solution, 0.05% Trypsin & 0.53mM EDTA, without Ca 2+ & Mg 2+ is available in a 100 mL format.
About This Item
Recommended Products
sterility
non-sterile
form
liquid
manufacturer/tradename
Specialty Media
input
sample type hematopoietic stem cell(s)
sample type: human embryonic stem cell(s)
sample type mesenchymal stem cell(s)
sample type induced pluripotent stem cell(s)
sample type cardiac stem cell(s)
sample type: mouse embryonic stem cell(s)
sample type epithelial cells
sample type neural stem cell(s)
sample type pancreatic stem cell(s)
Application
Storage and Stability
Disclaimer
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service